missing translation for 'onlineSavingsMsg'
Learn More

CC2D1A Antibody, Novus Biologicals™

Product Code. 18368859 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368859 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368859 Supplier Novus Biologicals Supplier No. H00054862B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

CC2D1A Polyclonal antibody specifically detects CC2D1A in Human, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CC2D1A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.4)
Gene Accession No. NP_060191.3
Gene Alias Akt kinase-interacting protein 1, coiled-coil and C2 domain containing 1A, coiled-coil and C2 domain-containing protein 1A, Five repressor element under dual repression-binding protein 1, FLJ20241, FLJ41160, FRE under dual repression-binding protein 1, Freud-1, mental retardation, nonsyndromic, autosomal recessive, 3, MRT3Freud-1/Aki1, Putative NF-kappa-B-activating protein 023N, putative NFkB activating protein
Host Species Mouse
Immunogen CC2D1A (NP_060191.3, 1 a.a. - 951 a.a.) full-length human protein. MHKRKGPPGPPGRGAAAARQLGLLVDLSPDGLMIPEDGANDEELEAEFLALVGGQPPALEKLKGKGPLPMEAIEKMASLCMRDPDEDEEEGTDEDDLEADDDLLAELNEVLGEEQKASETPPPVAQPKPEAPHPGLETTLQERLALYQTAIESARQAGDSAKMRRYDRGLKTLENLLASIRKGNAIDEADIPPPVAIGKGPASTPTYSPAPTQPAPRIASAPEPRVTLEGPSATAPASSPGLAKPQMPPGPCSPGPLAQLQSRQRDYKLAALHAKQQGDTTAAARHFRVAKSFDAVLEALSRGEPVDLSCLPPPPDQLPPDPPSPPSQPPTPATAPSTTEVPPPPRTLLEALEQRMERYQVAAAQAKSKGDQRKARMHERIVKQYQDAIRAHKAGRAVDVAELPVPPGFPPIQGLEATKPTQQSLVGVLETAMKLANQDEGPEDEEDEVPKKQNSPVAPTAQPKAPPSRTPQSGSAPTAKAPPKATSTRAQQQLAFLEGRKKQLLQAALRAKQKNDVEGAKMHLRQAKGLEPMLEASRNGLPVDITKVPPAPVNKDDFALVQRPGPGLSQEAARRYGELTKLIRQQHEMCLNHSNQFTQLGNITETTKFEKLAEDCKRSMDILKQAFVRGLPTPTARFEQRTFSVIKIFPDLSSNDMLLFIVKGINLPTPPGLSPGDLDVFVRFDFPYPNVEEAQKDKTSVIKNTDSPEFKEQFKLCINRSHRGFRRAIQTKGIKFEVVHKGGLFKTDRVLGTAQLKLDALEIACEVREILEVLDGRRPTGGRLEVMVRIREPLTAQQLETTTERWLVIDPVPAAVPTQVAGPKGKAPPVPAPARESGNRSARPLHSLSVLAFDQERLERKILALRQARRPVPPEVAQQYQDIMQRSQWQRAQLEQGGVGIRREYAAQLERQLQFYTEAARRLGNDGSRDAAKEALYRRNLVESELQRLRR
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54862
Target Species Human, Drosophila
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.