missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC2D1A Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33196-100ul
This item is not returnable.
View return policy
Description
CC2D1A Monoclonal antibody specifically detects CC2D1A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CC2D1A | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Akt kinase-interacting protein 1, coiled-coil and C2 domain containing 1A, coiled-coil and C2 domain-containing protein 1A, Five repressor element under dual repression-binding protein 1, FLJ20241, FLJ41160, FRE under dual repression-binding protein 1, Freud-1, mental retardation, nonsyndromic, autosomal recessive, 3, MRT3Freud-1/Aki1, Putative NF-kappa-B-activating protein 023N, putative NFkB activating protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 750-951 of human CC2D1A (Q6P1N0).,, Sequence:, VLGTAQLKLDALEIACEVREILEVLDGRRPTGGRLEVMVRIREPLTAQQLETTTERWLVIDPVPAAVPTQVAGPKGKAPPVPAPARESGNRSARPLHSLSVLAFDQERLERKILALRQARRPVPPEVAQQYQDIMQRSQWQRAQLEQGGVGIRREYAAQLERQLQFYTEAARRLGNDGSRDAAKEALYRRNLVESELQRLRR | |
| 100 μL | |
| DNA Repair, Signal Transduction | |
| 54862 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction