missing translation for 'onlineSavingsMsg'
Learn More

CC0651, MedChemExpress

Product Code. 16461499
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16461499

Brand: Medchem Express HY15301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CC0651 is an allosteric inhibitor of the human Cdc34 ubiquitin-conjugating enzyme. CC0651 potently (IC50=1.72 μM) inhibits the ubiquitination of p27Kip1, as confirmed by dose-response analysis.

CC0651 is an allosteric inhibitor of the human Cdc34 ubiquitin-conjugating enzyme. CC0651 potently (IC50=1.72 μM) inhibits the ubiquitination of p27Kip1, as confirmed by dose-response analysis.

Specifications

Chemical Name or Material CC0651
Physical Form Solid
CAS 1319207-44-7
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C20H21Cl2NO6
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 56 mg/mL (126.61 mM)
SMILES ClC1=CC(Cl)=CC(C2=CC=C(C=C2)C[C@@H]([C@@H]([C@H](C(O)=O)O)O)NC(COC)=O)=C1
Molecular Weight (g/mol) 442.29
Formula Weight 442.29
Percent Purity 98.44%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.