missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC Chemokine Receptor D6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57954-25ul
This item is not returnable.
View return policy
Description
CC Chemokine Receptor D6 Polyclonal specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CC Chemokine Receptor D6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| ACKR2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAATASPQPLATEDADSENSSFYYYDYLDEVAFMLCRKDAVVSFGKVFLP | |
| 25 μL | |
| Autophagy, Cellular Markers, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1238 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction