missing translation for 'onlineSavingsMsg'
Learn More

CC-90003, MedChemExpress

Product Code. 16477367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477367 5 mg 5mg
16475486 100 mg 100mg
16433186 25 mg 25mg
16404658 10 mg 10mg
16430947 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477367 Supplier Medchem Express Supplier No. HY112570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CC-90003 is an irreversible and selective inhibitor of ERK 1/2 with antitumor activity.

CC-90003 is an irreversible and selective inhibitor of ERK 1/2 with antitumor activity.

Specifications

Chemical Name or Material CC-90003
CAS 1621999-82-3
Color White
Purity Grade Notes Research
Molecular Formula C22H21F3N6O2
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (272.66 mM)
SMILES C=CC(NC1=CC(C)=CC=C1NC2=NC(NC3=CC(OC)=NC=C3C)=NC=C2C(F)(F)F)=O
Molecular Weight (g/mol) 458.44
Formula Weight 458.44
Percent Purity 99.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.