missing translation for 'onlineSavingsMsg'
Learn More

CC-885, MedChemExpress

Product Code. 16473388
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473388 10 mg 10mg
16469916 50 mg 50mg
16454626 100 mg 100mg
16452746 25 mg 25mg
16436117 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473388 Supplier Medchem Express Supplier No. HY101488
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity.

CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity.

Specifications

Chemical Name or Material CC-885
CAS 1010100-07-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClN4O4
Quantity 10 mg
Solubility Information DMSO : 67.5 mg/mL (153.10 mM; Need ultrasonic)
SMILES O=C(NCC1=CC2=C(C(N(C(CC3)C(NC3=O)=O)C2)=O)C=C1)NC4=CC=C(C)C(Cl)=C4
Molecular Weight (g/mol) 440.88
Formula Weight 440.88
Percent Purity 99.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.