missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CC 401 dihydrochloride
GREENER_CHOICE

Product Code. 30001031
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001031

Brand: Tocris Bioscience™ 6258/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity JNK inhibitor; also inhibits HCMV replication

High affinity JNK inhibitor (Ki values are 25-50 nM). Inhibits JNK via competitive binding of the ATP-binding site of active, phosphorylated JNK. Exhibits > 40-fold selectivity for JNK over p38, ERK, IKK2, protein kinase C, Lck and ZAP70. Hepatoprotective. Also inhibits HCMV replication.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2250025-96-6

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Formula Weight 461.39
Physical Form Solid
Purity >98%
Chemical Name or Material 3-[3-[2-(1-Piperidinyl)ethoxy]phenyl]-5-(1H-1,2,4-triazol-5-yl)-1H-indazole dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.