missing translation for 'onlineSavingsMsg'
Learn More

Cbz-Protected R-(+)-Carbidopa, TRC

Product Code. 30387721
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387721 2.5 mg 2.50mg
30381262 5 mg 5mg
30375166 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387721 Supplier Toronto Research Chemicals Supplier No. TRCD4318712.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cbz-Protected R-(+)-Carbidopa
Analyte or Component Names Cbz-Protected R-(+)-Carbidopa
Shipping Condition Room Temperature
Molecular Formula C25 H34 N2 O6
Synonym Benzyl (R)-2-(3-(3,4-Dimethoxyphenyl)-2-methyl-1-oxo-1-(pentan-3-yloxy)propan-2-yl)hydrazine-1-carboxylate
InChI Formula InChI=1S/C25H34N2O6/c1-6-20(7-2)33-23(28)25(3,16-19-13-14-21(30-4)22(15-19)31-5)27-26-24(29)32-17-18-11-9-8-10-12-18/h8-15,20,27H,6-7,16-17H2,1-5H3,(H,26,29)/t25-/m1/s1
Solution Type Neat
SMILES CCC(CC)OC(=O)[C@@](C)(Cc1ccc(OC)c(OC)c1)NNC(=O)OCc2ccccc2
IUPAC Name pentan-3-yl (2R)-3-(3,4-dimethoxyphenyl)-2-methyl-2-(2-phenylmethoxycarbonylhydrazinyl)propanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 458.547
Quantity 2.5 mg
Formula Weight 458.242
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.