missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38232-100ul
This item is not returnable.
View return policy
Description
Cbx8 Polyclonal antibody specifically detects Cbx8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Cbx8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| chromobox homolog 8, chromobox homolog 8 (Drosophila Pc class), chromobox homolog 8 (Pc class homolog, Drosophila), hPc3, Pc3, PC3chromobox protein homolog 8, polycomb 3, Polycomb 3 homolog, RC1Pc class 3 homolog, Rectachrome 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-280 of human Cbx8 (NP_065700.1).,, Sequence:, LLKAQAKAKAKTYEFRSDSARGIRIPYPGRSPQDLASTSRAREGLRNMGLSPPASSTSTSSTCRAEAPRDRDRDRDRDRERDRERERERERERERERERERGTSRVDDKPSSPGDSSKKRGPKPRKELPDPSQRPLGEPSAGLGEYLKGRKLDDTPSGAGKFPAGHSVIQLARRQDSDLVQCGVTSPSSAEATGKLAVDTF | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57332 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction