Learn More
Invitrogen™ CBX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596352
Description
Immunogen sequence: AVGLNLSHVR KRCLSETHGE REPCKKRLTA RSISTPTCLG GSPAAERPAD LPPAAALPQP EVILLDSDLD EPIDLRCVKT RSEAGEPPSS LQVKPETPAS AAVAVAAAAA PTTTAEKPPA EAQDEPAESL SEFKPFFGNI IITDVTANCL TVTFKEYVTV; Positive Samples: MCF-7, HepG2, SW620, Jurkat, Mouse brain, Mouse liver, Rat liver; Cellular Location: Nucleus, Nucleus speckle.
Polycomb group (PcG) proteins are chromatin-associated proteins which are important for embryonic and adult stem cell self-renewal and maintenance. At least two distinct human PcG complexes have been identified. Polycomb repressive complex I (PRC1) contains CBX4/Pc2, HPH1 and RING domain-containing proteins (RING1, RING2 and BMI1). CBX4 (Chromobox protein homolog 4 or PC2), which functions as a transcriptional suppressor, is a candidate of KyoT2-binding proteins. It is a E3 SUMO-protein ligase which facilitates SUMO1 conjugation. Depletion of CBX4 results in decreased cellular resistance to ionizing radiation. CBX4 is directly involved in the cellular response to DNA damage.
Specifications
| CBX4 | |
| Polyclonal | |
| Unconjugated | |
| CBX4 | |
| Cbx4; chromobox 4; chromobox homolog 4; chromobox homolog 4 (Pc class homolog, Drosophila); chromobox protein homolog 4; chromobox-like protein 4; E3 SUMO-protein ligase CBX4; E3 SUMO-protein transferase CBX4; hPc2; MPc2; NBP16; NS5ATP1-binding protein 16; Pc class 2 homolog; PC2; Polycomb 2 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12418, 501403, 8535 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00257, O55187 | |
| CBX4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 401-560 of human CBX4 (NP_0036462). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.