missing translation for 'onlineSavingsMsg'
Learn More

CBX4 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18428646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428646 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428646 Supplier Novus Biologicals Supplier No. NBP281972MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CBX4 Polyclonal antibody specifically detects CBX4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX4
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog
Host Species Rabbit
Immunogen CBX4 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human CBX4. The immunogen is located within amino acids 50 - 100 of CBX4. Amino Acid Squence: EQLMGYRKRGPKPKPLV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8535
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.