missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £549.00
Specifications
| Antigen | CBX3 |
|---|---|
| Dilution | ELISA, Dot Blot 1:500 - 1:1000 |
| Applications | ELISA, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232334
|
Novus Biologicals
NBP3-35891-100ul |
100 μL |
£549.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228937
|
Novus Biologicals
NBP3-35891-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBX3 Polyclonal antibody specifically detects CBX3 in Human samples. It is validated for ELISA,Dot BlotSpecifications
| CBX3 | |
| ELISA, Dot Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 11335 | |
| IgG | |
| Affinity purified |
| ELISA, Dot Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox protein homolog 3, Drosophila), HECH, Heterochromatin protein 1 homolog gamma, heterochromatin protein HP1 gamma, heterochromatin-like protein 1, HP1 gamma, HP1 gamma homolog, HP1-GAMMA, Modifier 2 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-183 of human CBX3 (NP_009207.2).,, Sequence:, MASNKTTLQKMGKKQNGKSKKVEEAEPEEFVVEKVLDRRVVNGKVEYFLKWKGFTDADNTWEPEENLDCPELIEAFLNSQKAGKEKDGTKRKSLSDSESDDSKSKKKRDAADKPRGFARGLDPERIIGATDSSGELMFLMKWKDSDEADLVLAKEANMKCPQIVIAFYEERLTWHSCPEDEAQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title