missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBX2 Recombinant Human Monoclonal Antibody (RAB-C238)

Human Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA551922

Product Code. 30087693

  • £614.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody recognizes CBX2 (Chromobox protein homolog 2). It binds to a folded domain, amino acids 2-70. CBX2 is a component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development.

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CBX2
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Q14781
CBX2
This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX2 protein under non-denaturing conditions.
100 μg
Primary
Human
Antibody
F(ab) κ
ChIP Assay, ELISA, Immunocytochemistry, Mass Spectrometry
RAB-C238
Unconjugated
CBX2
Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5
Human
Metal-chelate chromatography
RUO
84733
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.