missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbx2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-40342
This item is not returnable.
View return policy
Description
Cbx2 Polyclonal specifically detects Cbx2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative), Chromatin Immunoprecipitation Sequencing.
Specifications
| Cbx2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CBX2 | |
| The immunogen for this product maps to a region between residue 482 and 532 of human chromobox homolog 2 using the numbering given in entry NP_005180.1 (GeneID 84733). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| ChIP Assay, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-10 ug/mg lysate, Western Blot (Negative), Chromatin Immunoprecipitation Sequencing 1 ug | |
| CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84733 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction