missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10241-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
| CBX1 | |
| Polyclonal | |
| Unconjugated | |
| CBX1 | |
| CBX, CBX1, Chromobox protein homolog 1, Heterochromatin protein p25, HP1 BETA, HP1Hs beta, HP1Hsbeta, M31, MOD1, Modifier 1 protein, p25beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10951, 12412, 360609 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P83916, P83917 | |
| Cbx1 | |
| CBX1 Fusion Protein Ag0328 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction