Learn More
CBX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10241-2-AP-150UL
 View more versions of this product
Product Code. 16886493
  • £349.00 / 150µL
Description
This gene encodes a highly conserved nonhistone protein, which is a member of the heterochromatin protein family. The protein is enriched in the heterochromatin and associated with centromeres. The protein has a single N-terminal chromodomain which can bind to histone proteins via methylated lysine residues, and a C-terminal chromo shadow-domain which is responsible for the homodimerization and interaction with a number of chromatin-associated nonhistone proteins. The protein may play an important role in the epigenetic control of chromatin structure and gene expression. Several related pseudogenes are located on chromosomes 1, 3, and X. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
Specifications
CBX1
Polyclonal
Unconjugated
CBX1
CBX, CBX1, Chromobox protein homolog 1, Heterochromatin protein p25, HP1 BETA, HP1Hs beta, HP1Hsbeta, M31, MOD1, Modifier 1 protein, p25beta
Rabbit
Antigen Affinity Chromatography
RUO
10951, 12412, 360609
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P83916, P83917
Cbx1
CBX1 Fusion Protein Ag0328
150 μL
Primary
Human, Mouse, Rat