missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBWD1 Antibody (OTI4D4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72027PCP
This item is not returnable.
View return policy
Description
CBWD1 Monoclonal specifically detects CBWD1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| CBWD1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CBWD1 | |
| Full length recombinant protein of human CBWD1 (NP_060961) produced in HEK293T cells. | |
| 0.1 mL | |
| 55871.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4D4 | |
| Flow Cytometry | |
| Cobalamin synthase W domain-containing protein 1, cobalamin synthetase W domain-containing protein 1, COBP, COBW domain containing 1, COBW domain-containing protein 1, COBW-like protein, NPC-A-6, NPC-A-6 COBW domain-containing protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat, Canine, Primate, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction