missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532074
This item is not returnable.
View return policy
Description
Recommended positive controls: Raji. Predicted reactivity: Mouse (85%), Rat (83%), Rhesus Monkey (97%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The heterotetramer with HSD17B8 has NADH-dependent 3-ketoacyl-acyl carrier protein reductase activity, and thereby plays a role in mitochondrial fatty acid biosynthesis. Within the heterotetramer, HSD17B8 binds NADH; CBR4 binds NADPD. The homotetramer has NADPH-dependent quinone reductase activity. Both homotetramer and the heterotetramer have broad substrate specificity and can reduce 9,10-phenanthrenequinone, 1,4-benzoquinone and various other o-quinones and p-quinones (in vitro).
Specifications
| CBR4 | |
| Polyclonal | |
| Unconjugated | |
| CBR4 | |
| 3-ketoacyl-[acyl-carrier-protein] reductase beta subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; A730083J17Rik; BC009118; carbonic reductase 4; carbonyl reductase 4; carbonyl reductase family member 4; Cbr4; KAR beta subunit; Quinone reductase CBR4; SDR45C1; Short chain dehydrogenase/reductase family 45C member 1; short chain dehydrogenase/reductase family 45C, member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N4T8 | |
| CBR4 | |
| Recombinant protein encompassing a sequence within the center region of human CBR4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction