missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR3 Mouse anti-Human, Mouse, Clone: 7.00E+08, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59401-100UL
This item is not returnable.
View return policy
Description
CBR3 Monoclonal specifically detects CBR3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CBR3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1013, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 700000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| CBR3 | |
| Recombinant human CBR3 (1-277aa) purified from E. coli | |
| 100 μL | |
| Lipid and Metabolism | |
| 874 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction