missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBR3 Monoclonal Antibody (OTI1G6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525626
This item is not returnable.
View return policy
Description
CBR3 Monoclonal Antibody for Western Blot, IHC (P)
Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
Specifications
| CBR3 | |
| Monoclonal | |
| 0.43 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75828, Q8K354 | |
| CBR3 | |
| Full length human recombinant protein of CBR3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G6 | |
| Unconjugated | |
| CBR3 | |
| 1110001J05Rik; C81353; carbonyl reductase (NADPH) 3; carbonyl reductase [NADPH] 3; carbonyl reductase 3; Cbr3; epididymis secretory protein Li 25; hCBR3; HEL-S-25; NADPH-dependent carbonyl reductase 3; SDR21C2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 109857, 304078, 487748, 874 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction