missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR3 Antibody (S06-7B3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19596-100ul
This item is not returnable.
View return policy
Description
CBR3 Monoclonal antibody specifically detects CBR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| CBR3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 10 μg/ml, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 3 μg/ml, Sandwich ELISA 1:100-1:2000 | |
| carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2 | |
| A synthetic peptide of human CBR3 (Uniprot # O75828) | |
| 100 μg | |
| Lipid and Metabolism | |
| 874 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| S06-7B3 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction