missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £367.00
Specifications
| Antigen | CBR1 |
|---|---|
| Applications | Western Blot, Immunofluorescence, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18364056
|
Novus Biologicals
NBP3-03418-20UL |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18353127
|
Novus Biologicals
NBP3-03418-100UL |
100 μL |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBR1 Polyclonal specifically detects CBR1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| CBR1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human CBR1 (NP_001748.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 873 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title