missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR1 Antibody (4E12), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74041V
This item is not returnable.
View return policy
Description
CBR1 Monoclonal specifically detects CBR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CBR1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CBR1 | |
| Recombinant human CBR1 (1-277 aa ) purified from E. coli | |
| 100 μL | |
| Stem Cell Markers | |
| 873 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4000000000000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human CBR1 mAb, clone 4E12, is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CBR1 protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction