Learn More
CBP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11950-1-AP-20UL
 View more versions of this product
Product Code. 16899353
  • £135.00 / 20µL
Description
The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5′ cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3′-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CBP20
Polyclonal
Unconjugated
NCBP2
CBC2, CBP20, NCBP 20 kDa subunit, NCBP interacting protein 1, NCBP2, NIP1, PIG55
Rabbit
Antigen Affinity Chromatography
RUO
22916, 68092, 689116
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B1WC40, P52298, Q9CQ49
Ncbp2
CBP20 Fusion Protein Ag2550
20 μL
Primary
Human, Mouse, Rat