missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11950-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is a component of the nuclear cap-binding protein complex (CBC), which binds to the monomethylated 5′ cap of nascent pre-mRNA in the nucleoplasm. The encoded protein has an RNP domain commonly found in RNA binding proteins, and contains the cap-binding activity. The CBC promotes pre-mRNA splicing, 3′-end processing, RNA nuclear export, and nonsense-mediated mRNA decay. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| CBP20 | |
| Polyclonal | |
| Unconjugated | |
| NCBP2 | |
| CBC2, CBP20, NCBP 20 kDa subunit, NCBP interacting protein 1, NCBP2, NIP1, PIG55 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22916, 68092, 689116 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B1WC40, P52298, Q9CQ49 | |
| Ncbp2 | |
| CBP20 Fusion Protein Ag2550 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction