missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBP20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CBP20 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18239754
|
Novus Biologicals
NBP2-56303 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637218
|
Novus Biologicals
NBP2-56303-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBP20 Polyclonal specifically detects CBP20 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| CBP20 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 22916 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GQVRDEYRQDYDAGRGGYGKLAQNQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 20 kDa nuclear cap-binding protein, Cbc2, CBP20CBC2, NCBP 20 kDa subunit, NCBP interacting protein 1, NCBP-interacting protein 1, NIP1Cell proliferation-inducing gene 55 protein, nuclear cap binding protein subunit 2, 20kD, nuclear cap binding protein subunit 2, 20kDa, nuclear cap-binding protein subunit 2 | |
| NCBP2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title