missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBP Recombinant Rabbit Monoclonal Antibody (HL2197)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550428
This item is not returnable.
View return policy
Description
CBP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
Specifications
| CBP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P45481, Q6JHU9, Q92793 | |
| CREBBP | |
| Recombinant fragmemt of human CBP. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2197 | |
| Unconjugated | |
| CREBBP | |
| AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 12914, 1387, 54244 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction