Learn More
Invitrogen™ CBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532367
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
Specifications
| CBP | |
| Polyclonal | |
| PBS with 1% BSA and <0.1% sodium azide; pH 7.6 | |
| Q92793 | |
| CREBBP | |
| Synthetic peptide corresponding to C-terminus of human CBP. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| CREBBP | |
| AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1387 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.