missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calmodulin binding peptide Recombinant Rabbit Monoclonal Antibody (HL1841)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547197
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
CBP affinity tag is a useful tag in protein study which can be added to the N or C terminus of proteins of interest through DNA recombinant technology. The tag is derived from muscle myosin light-chain kinase. The tag comprises 26 amino acid residues with the molecular weight of 4 kDa and sequence of KRRWKKNFIAVSAANRFKKISSSGAL. CBP tag has the relatively high affinity for calmodulin (CaM). CBP-tagged proteins can be purified from crude cell extracts through CaM affinity resin. CBP Tag antibody is a useful tool in analysis of CBP-tagged proteins.
Specifications
| Calmodulin binding peptide | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1841 | |
| Unconjugated | |
| AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS | |
| Synthetic peptide of human CBP tag. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction