missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBP/KAT3A Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-381IR
This item is not returnable.
View return policy
Description
CBP/KAT3A Polyclonal antibody specifically detects CBP/KAT3A in Human samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP)
Specifications
| CBP/KAT3A | |
| Polyclonal | |
| Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
| CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome | |
| The immunogen recognized by this antibody maps to a region between residues 975 and 1025 of human CREB Binding Protein using the numbering given in entry NP_004371.1 (GeneID 1387). | |
| 0.1 mL | |
| Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 1387 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation, ChIP Assay | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction