missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBLB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D3A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66353-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.Specifications
| CBLB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13191, Q3TTA7, Q8K4S7 | |
| Cblb | |
| CBLB Fusion Protein Ag18757 | |
| 20 μL | |
| Primary | |
| Pig, Human, Rat, Mouse | |
| Antibody | |
| IgG3 |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 2D3A2 | |
| Unconjugated | |
| CBLB | |
| CBLB, Nbla00127, RING finger protein 56, RNF56, SH3 binding protein CBL B | |
| Mouse | |
| Protein A | |
| RUO | |
| 100621690, 171136, 208650, 868 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction