missing translation for 'onlineSavingsMsg'
Learn More

CBLB Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18416542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416542 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416542 Supplier Novus Biologicals Supplier No. NBP149920AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CBLB Polyclonal antibody specifically detects CBLB in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBLB
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Cas-Br-M (murine) ecotropic retroviral transforming sequence b, Cas-Br-M (murine) ectropic retroviral transforming sequence b, Casitas B-lineage lymphoma proto-oncogene b, DKFZp686J10223, DKFZp779A0729, DKFZp779F1443, E3 ubiquitin-protein ligase CBL-B, EC 6.3.2, EC 6.3.2.-, FLJ41152, RING finger protein 56, RNF56FLJ36865, SH3-binding protein CBL-B, Signal transduction protein CBL-B
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 932 and 982 of human Cas-Br-M (murine) Ecotropic Retroviral Transforming Sequence b using the numbering given in entry NP_733762.2 (GeneID 868).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 868
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.