missing translation for 'onlineSavingsMsg'
Learn More

CBL0137 hydrochloride, MedChemExpress

Product Code. 16470306
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470306 2 mg 2mg
16463616 25 mg 25mg
16453657 50 mg 50mg
16437648 10 mg 10mg
16410118 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470306 Supplier Medchem Express Supplier No. HY18935A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CBL0137 hydrochloride is an inhibitor of the histone chaperone, FACT. CBL0137 hydrochloride can also activate p53 and inhibits NF-κB with EC50s of 0.37 and 0.47 μM, respectively.

CBL0137 hydrochloride is an inhibitor of the histone chaperone, FACT. CBL0137 hydrochloride can also activate p53 and inhibits NF-κB with EC50s of 0.37 and 0.47 μM, respectively.

Specifications

Chemical Name or Material CBL0137 hydrochloride
CAS 1197397-89-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25ClN2O2
Quantity 2 mg
Synonym Curaxin-137 hydrochloride CBL-C137 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (89.38 mM; Need ultrasonic) ∣H2O : 10 mg/mL (26.82 mM; Need ultrasonic)
SMILES CC(NCCN1C2=C(C3=C1C=CC(C(C)=O)=C3)C=C(C(C)=O)C=C2)C.[H]Cl
Molecular Weight (g/mol) 372.89
Formula Weight 372.89
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.