missing translation for 'onlineSavingsMsg'
Learn More

CBL0137 hydrochloride, MedChemExpress

Product Code. 16437648
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437648

Brand: Medchem Express HY18935A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CBL0137 hydrochloride is an inhibitor of the histone chaperone, FACT. CBL0137 hydrochloride can also activate p53 and inhibits NF-κB with EC50s of 0.37 and 0.47 μM, respectively.

CBL0137 hydrochloride is an inhibitor of the histone chaperone, FACT. CBL0137 hydrochloride can also activate p53 and inhibits NF-κB with EC50s of 0.37 and 0.47 μM, respectively.

Specifications

Chemical Name or Material CBL0137 hydrochloride
CAS 1197397-89-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H25ClN2O2
Quantity 10 mg
Synonym Curaxin-137 hydrochloride CBL-C137 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (89.38 mM; Need ultrasonic) ∣H2O : 10 mg/mL (26.82 mM; Need ultrasonic)
SMILES CC(NCCN1C2=C(C3=C1C=CC(C(C)=O)=C3)C=C(C(C)=O)C=C2)C.[H]Cl
Molecular Weight (g/mol) 372.89
Formula Weight 372.89
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.