missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbl-c Antibody (OTI3B4), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71755PECY7
This item is not returnable.
View return policy
Description
Cbl-c Monoclonal specifically detects Cbl-c in Human samples. It is validated for Flow Cytometry.
Specifications
| Cbl-c | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| CBLC | |
| Recombinant protein fragment corresponding to amino acids 244-474 of human Cbl-c (NP_036248) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| OTI3B4 | |
| Flow Cytometry | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 23624.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction