missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbl-c Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97089-50ul
This item is not returnable.
View return policy
Description
Cbl-c Polyclonal antibody specifically detects Cbl-c in Human samples. It is validated for ELISA
Specifications
| Cbl-c | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
| Produced in rabbits immunized with purified, recombinant Human Cbl-c (Accession#: AAH28915.1; Met1-Ala474) | |
| 50 μg | |
| Stem Cell Markers | |
| 23624 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction