missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cbl-c Antibody (01), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06621-50ul
This item is not returnable.
View return policy
Description
Cbl-c Monoclonal antibody specifically detects Cbl-c in Human samples. It is validated for ELISA
Specifications
| Cbl-c | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 01 | |
| ELISA 1:1000-1:2000 | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cbl-c (Accession#: AAH28915.1; Met1-Ala474). | |
| 50 μg | |
| Stem Cell Markers | |
| 23624 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction