Learn More
Cbl-b Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA554107
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DNRLSRHIHH VESVPSRDPP MPLEAWCPRD VFGTNQLVGC RLLGEGSPKP GITASSNVNG RHSRVGSDPV LMRKHRRHDL PLEGAKVFSN GHLGSEEYDV PPRLSPPPPV TTLLPSIKCT GPLA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.Specifications
Cbl-b | |
Polyclonal | |
Unconjugated | |
CBLB | |
AI429560; AI851073; Cas-Br-M (murine) ecotropic retroviral transforming sequence b; Cas-Br-M (murine) ectropic retroviral transforming sequence b; Casitas B-lineage lymphoma b; casitas B-lineage lymphoma proto-oncogene b; Cbl proto-oncogene B; Cbl proto-oncogene B, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase B; CBLB; Cbl-b; E3 ubiquitin-protein ligase CBL-B; Nbla00127; RING finger protein 56; RING-type E3 ubiquitin transferase CBL-B; RNF56; SH3-binding protein CBL-B; signal transduction protein CBL-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
868 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13191 | |
CBLB | |
Recombinant protein corresponding to Human Cbl-b | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only