missing translation for 'onlineSavingsMsg'
Learn More

CBFB Antibody (PCRP-CBFB-1E6), DyLight 755, Novus Biologicals™

Product Code. 18334204 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334204 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334204 Supplier Novus Biologicals Supplier No. NBP314059IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CBFB Monoclonal antibody specifically detects CBFB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBFB
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-CBFB-1E6
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CBF-beta, core-binding factor subunit beta, core-binding factor, beta subunit, PEA2-beta, PEBP2B, PEBP2-beta, polyomavirus enhancer binding protein 2, beta subunit, Polyomavirus enhancer-binding protein 2 beta subunit, SL3/AKV core-binding factor beta subunit, SL3-3 enhancer factor 1 beta subunit, SL3-3 enhancer factor 1 subunit beta
Host Species Mouse
Immunogen Recombinant full-length human CBFB protein (Uniprot: Q13951)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 865
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.