missing translation for 'onlineSavingsMsg'
Learn More

CBF1-interacting corepressor Antibody, DyLight 405, Novus Biologicals™

Product Code. 18425633 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425633 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425633 Supplier Novus Biologicals Supplier No. NBP171896V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CBF1-interacting corepressor Polyclonal antibody specifically detects CBF1-interacting corepressor in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBF1-interacting corepressor
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CBF1 interacting corepressor, CBF1-interacting corepressor, CIRrecepin, corepressor interacting with RBPJ 1, corepressor interacting with RBPJ, 1, Recepin
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human Corepressor Interacting with RBPJ 1 using the numbering given in entry NP_004873.3 (GeneID 9541).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 9541
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.