missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CBF beta Recombinant Rabbit Monoclonal Antibody (23GB3785), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CBF beta |
|---|---|
| Clone | 23GB3785 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087689
![]() |
Invitrogen™
MA553454 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087706
|
Invitrogen™
MA553455 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBF beta Recombinant Monoclonal antibody specifically detects CBF beta in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| CBF beta | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q08024, Q13951 | |
| 12400, 361391, 865 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3785 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI893578; CBFB; CBF-beta; core binding factor beta; core-binding factor subunit beta; core-binding factor, beta subunit; PEA2; PEA2-beta; Pebp2; PEBP2B; PEBP2-beta; Pebpb2; polyomavirus enhancer binding protein 2, beta subunit; polyomavirus enhancer-binding protein 2 beta subunit; SL3/AKV core-binding factor beta subunit; SL3-3 enhancer factor 1 beta subunit; SL3-3 enhancer factor 1 subunit beta | |
| A synthetic peptide of human CBFb (100-182AA). | |
| Primary | |
| -20°C | |
| CBFB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title