missing translation for 'onlineSavingsMsg'
Learn More

CBB1007 hydrochloride, MedChemExpress

Product Code. 16436448
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436448

Brand: Medchem Express HY15313B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CBB1007 Hcl is a cell-permeable amidino-guanidinium compound that acts as a potent, reversible and substrate competitive LSD1 selective inhibitor (IC50 = 5.27 μM for hLSD1).

CBB1007 Hcl is a cell-permeable amidino-guanidinium compound that acts as a potent, reversible and substrate competitive LSD1 selective inhibitor (IC50 = 5.27 μM for hLSD1).

Specifications

Chemical Name or Material CBB1007 hydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C27H39Cl5N8O4
Quantity 10 mg
SMILES [H]Cl.[H]Cl.[H]Cl.[H]Cl.[H]Cl.NC(N1CCN(CC1)CC2=CC(C(OC)=O)=CC(C(N3CCN(CC3)C(C4=CC=C(C=C4)C(N)=N)=O)=O)=C2)=N
Molecular Weight (g/mol) 716.91
Formula Weight 716.91
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.