missing translation for 'onlineSavingsMsg'
Learn More

CB1-IN-1, MedChemExpress

Product Code. 16495976
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495976

Brand: Medchem Express HY12790

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CB1-IN-1 (BPRCB1184) is a peripherally restricted CB1R antagonist, with Ki of 0.3 nM and 21 nM for CB1R (EC50 = 3 nM) and CB2R, respectively.

CB1-IN-1 (BPRCB1184) is a peripherally restricted CB1R antagonist, with Ki of 0.3 nM and 21 nM for CB1R (EC50 = 3 nM) and CB2R, respectively.

Specifications

Chemical Name or Material CB1-IN-1
Physical Form Solid
CAS 1429239-98-4
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C33H31Cl2F3N6O3S2
For Use With (Application) Neuroscience-Neuromodulation
Synonym BPRCB1184
Solubility Information DMSO : 125 mg/mL (166.30 mM; Need ultrasonic)
SMILES ClC1=CC(Cl)=CC=C1N2C(C3=CC=C(C#CC4=CC=C(C(F)(F)F)C=C4)S3)=C(CNS(=O)(N5CCCC5)=O)C(C(NN6CCCCC6)=O)=N2
Molecular Weight (g/mol) 751.67
Formula Weight 751.67
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.