missing translation for 'onlineSavingsMsg'
Learn More

CAY10650, MedChemExpress

Product Code. 16484168
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484168 10 mg 10mg
16470517 50 mg 50mg
16461099 1 mg 1mg
16446877 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484168 Supplier Medchem Express Supplier No. HY10801
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CAY10650 is a highly potent cytosolic phospholipase A2α (cPLA2α) inhibitor with an IC50 value of 12 nM. CAY10650 suppresses lipid droplets formation and PGE2 secretion.

CAY10650 is a highly potent cytosolic phospholipase A2α (cPLA2α) inhibitor with an IC50 value of 12 nM. CAY10650 suppresses lipid droplets formation and PGE2 secretion.

Specifications

Chemical Name or Material CAY10650
CAS 1233706-88-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H25NO6
Quantity 10 mg
Solubility Information DMSO : ≥ 43 mg/mL (91.20 mM)
SMILES O=C(CN1C(C=CC(C(O)=O)=C2)=C2C(C(C(C)C)=O)=C1)COC(C=C3)=CC=C3OC4=CC=CC=C4
Molecular Weight (g/mol) 471.5
Formula Weight 471.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.