missing translation for 'onlineSavingsMsg'
Learn More

CAY10650, MedChemExpress

Product Code. 16446877
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446877

Brand: Medchem Express HY10801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAY10650 is a highly potent cytosolic phospholipase A2α (cPLA2α) inhibitor with an IC50 value of 12 nM. CAY10650 suppresses lipid droplets formation and PGE2 secretion.

CAY10650 is a highly potent cytosolic phospholipase A2α (cPLA2α) inhibitor with an IC50 value of 12 nM. CAY10650 suppresses lipid droplets formation and PGE2 secretion.

Specifications

Chemical Name or Material CAY10650
CAS 1233706-88-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H25NO6
Quantity 5 mg
Solubility Information DMSO : ≥ 43 mg/mL (91.20 mM)
SMILES O=C(CN1C(C=CC(C(O)=O)=C2)=C2C(C(C(C)C)=O)=C1)COC(C=C3)=CC=C3OC4=CC=CC=C4
Molecular Weight (g/mol) 471.5
Formula Weight 471.5
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.