missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CAY10603
Description
- CAY10603 is a selective and potent inhibitor of histone deacetylase 6 (HDAC6; IC50 = 0.002 nM; Kozikowski et al.) HDAC6 deacetylates lysine residues on the N-terminal part of histones
- HDAC6 is also known to regulate heat shock protein 90 (hsp90) via hyperacetylation (Rao et al.)
- Inhibits growth of several pancreatic cancer cell lines (IC50 = 0.1 - 1 μM) and SCCOHT, an ovarian cancer cell line (Kozikowski et al.; Wang et al.)
Specifications
Specifications
| CAS | 1045792-66-2 |
| Synonym | HDAC6 inhibitor |
| Quantity | 1 mg |
| Molecular Formula | C22H30N4O6 |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?