missing translation for 'onlineSavingsMsg'
Learn More

CAY10603, MedChemExpress

Product Code. 16473557
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473557 50 mg 50mg
16457538 10 mg 10mg
16453606 25 mg 25mg
16447486 100 mg 100mg
16441739 1 mg 1mg
16440018 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16473557 Supplier Medchem Express Supplier No. HY18613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM.

CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM

Specifications

Chemical Name or Material CAY10603
Physical Form Solid
CAS 1045792-66-2
Color Light Nude
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H30N4O6
For Use With (Application) Cancer-programmed cell death
Synonym BML-281
Solubility Information DMSO : ≥ 50 mg/mL (111.98 mM)
SMILES O=C(OC(C)(C)C)NC1=CC=C(C2=CC(C(NCCCCCCC(NO)=O)=O)=NO2)C=C1
Molecular Weight (g/mol) 446.5
Formula Weight 446.5
Percent Purity 95.39%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.