missing translation for 'onlineSavingsMsg'
Learn More

CAY10602, MedChemExpress

Product Code. 16494966
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494966 100 mg 100mg
16486667 5 mg 5mg
16460307 50 mg 50mg
16403958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494966 Supplier Medchem Express Supplier No. HY104073

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAY10602 is a SIRT1 activator. CAY10602 dose-dependently suppresses the NF-κB-dependent induction of TNF-α by lipopolysaccharide in THP-1 cells.

CAY10602 is a SIRT1 activator. CAY10602 dose-dependently suppresses the NF-κB-dependent induction of TNF-α by lipopolysaccharide in THP-1 cells.

Specifications

Chemical Name or Material CAY10602
CAS 374922-43-7
Color Gray
Purity Grade Notes Research
Molecular Formula C22H15FN4O2S
Quantity 100 mg
Solubility Information DMSO : 41.67 mg/mL (99.58 mM; Need ultrasonic)
SMILES NC1=C(S(=O)(C2=CC=CC=C2)=O)C3=NC4=CC=CC=C4N=C3N1C5=CC=C(F)C=C5
Molecular Weight (g/mol) 418.44
Formula Weight 418.44
Percent Purity 98.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.