missing translation for 'onlineSavingsMsg'
Learn More

CAY10505, MedChemExpress

Product Code. 16441897
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16441897

Brand: Medchem Express HY13530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons.

CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons.

Specifications

Chemical Name or Material CAY10505
CAS 1218777-13-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8FNO3S
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (117.53 mM)
SMILES O=C(NC/1=O)SC1=C\C2=CC=C(C3=CC=C(F)C=C3)O2
Molecular Weight (g/mol) 289.28
Formula Weight 289.28
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.