missing translation for 'onlineSavingsMsg'
Learn More

CAY10505, MedChemExpress

Product Code. 16463366
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463366 25 mg 25mg
16456216 100 mg 100mg
16441897 50 mg 50mg
16425738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463366 Supplier Medchem Express Supplier No. HY13530
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons.

CAY10505 is a potent and selective PI3Kγ inhibitor with an IC50 of 30 nM in neurons.

Specifications

Chemical Name or Material CAY10505
CAS 1218777-13-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8FNO3S
Quantity 25 mg
Solubility Information DMSO : ≥ 34 mg/mL (117.53 mM)
SMILES O=C(NC/1=O)SC1=C\C2=CC=C(C3=CC=C(F)C=C3)O2
Molecular Weight (g/mol) 289.28
Formula Weight 289.28
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.